Learn More
Medchemexpress LLC HY-P1071A 1mg Medchemexpress, α-CGRP(human) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1071A1MG
Encompass_Preferred
Medchemexpress, HY-P1071A 1mg α-CGRP(human) (TFA) CAS: α-CGRP(human) TFA is a regulatory neuropeptide of 37 amino acids. α-CGRP(human) is widely distributed in the central and peripheral nervous system. α-CGRP(human) TFA is a potent vasodilator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3037