Learn More
Aviva Systems Biology Corp CGRRF1 Antibody - middle region (ARP43075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CGRRF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CGRRF1. Alternate names: CGR19, RNF197ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53990