Learn More
TARGETMOL CHEMICALS INC CGS 15435 5MG
SDP
Catalog No. 502581665
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGS 15435 is a potent thromboxen (TxA2) synthetase inhibitor (IC50 1 nM). CGS 15435 acts on PGI2 synthase cyclooxygenase and liposynthase with much less selectivity than on thromboxin synthase. purity: 99%
Catalog No. 50-258-1665 Supplier TARGETMOL CHEMICALS INC Supplier No. T107835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CGS 15435 is a potent thromboxen (TxA2) synthetase inhibitor (IC50 1 nM). CGS 15435 acts on PGI2 synthase cyclooxygenase and liposynthase with much less selectivity than on thromboxin synthase. purity: 99%