Learn More
Selleck Chemical LLC CGS 21680 HCl 5mg
SDP
Catalog No. 502173079
Encompass_Preferred
CGS 21680 HCl is an adenosine A2 receptor agonist with IC50 of 22 nM, exhibits 140-fold over A1 receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3079 Supplier Selleck Chemical LLC Supplier No. S21535MG
CGS 21680 HCl is an adenosine A2 receptor agonist with IC50 of 22 nM, exhibits 140-fold over A1 receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.