Learn More
Medchemexpress LLC CGS 21680 hydrochloride | 124431-80-7 | MFCD00078573 | 99.7% | C23H30ClN7O6 | 10MG
SDP
Catalog No. 5000261013
Encompass_Preferred
CGS 21680 hydrochloride is a research-grade selective adenosine A2A receptor agonist (Ki = 27 nM) supplied as the hydrochloride salt for pharmacology and receptor-signaling studies.
  • Selective adenosine A2A receptor agonist with a Ki of 27 nM.
  • High reported purity (99.7%) suitable for reproducible assays.
  • Supplied as the hydrochloride salt to limit free-base instability.
  • Suitable for in vitro pharmacology and receptor-binding studies.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-61013 Supplier Medchemexpress LLC Supplier No. HY13201A10MG
CGS 21680 hydrochloride is a research-grade selective adenosine A2A receptor agonist (Ki = 27 nM) supplied as the hydrochloride salt for pharmacology and receptor-signaling studies.
  • Selective adenosine A2A receptor agonist with a Ki of 27 nM.
  • High reported purity (99.7%) suitable for reproducible assays.
  • Supplied as the hydrochloride salt to limit free-base instability.
  • Suitable for in vitro pharmacology and receptor-binding studies.
  • Available in small research pack sizes for laboratory use.