Learn More
Cayman Chemical CGS 21680 (hydrochloride), Purity: ≥98%, CAS: 124431-80-7, 10 mg
SDP
Catalog No. 502967320
Encompass_Preferred
A potent, selective agonist of the adenosine A2A receptor (Ki = 11-46 nM); less effective at adenosine A1 and A3 receptors (Kis = 0.5-3.1 and 0.6-1 µM, respectively) and is without effect at adenosine A2B (Ki > 10 µM)
Catalog No. 50-296-7320 Supplier Cayman Chemical Supplier No. 1712610 MG
A potent, selective agonist of the adenosine A2A receptor (Ki = 11-46 nM); less effective at adenosine A1 and A3 receptors (Kis = 0.5-3.1 and 0.6-1 µM, respectively) and is without effect at adenosine A2B (Ki > 10 µM)