Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CH-0793076 TFA (TP3076 TFA) | 2740278-76-4 | 98.7% | 572.53 g·mol⁻¹ | C28H27F3N4O6 | 5 MG
SDP

Catalog No. 5000239827
Encompass_Preferred

CH-0793076 TFA is a camptothecin-like hexacyclic analog and the active metabolite of TP300 that functions as a DNA topoisomerase I inhibitor. It is supplied as a trifluoroacetate salt for research use and has demonstrated antiproliferative activity, including activity against cells expressing BCRP.

  • Inhibits DNA topoisomerase I (reported IC50 ≈ 2.3 μM).
  • Active metabolite of TP300.
  • Demonstrates antiproliferative activity in cellular assays.
  • Provided as a trifluoroacetate salt suitable for biochemical studies.
  • Molecular weight 572.53 g·mol⁻¹; chemical formula C28H27F3N4O6.

Catalog No. 50-002-39827 Supplier Medchemexpress LLC Supplier No. HY107096B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CH-0793076 TFA is a camptothecin-like hexacyclic analog and the active metabolite of TP300 that functions as a DNA topoisomerase I inhibitor. It is supplied as a trifluoroacetate salt for research use and has demonstrated antiproliferative activity, including activity against cells expressing BCRP.

  • Inhibits DNA topoisomerase I (reported IC50 ≈ 2.3 μM).
  • Active metabolite of TP300.
  • Demonstrates antiproliferative activity in cellular assays.
  • Provided as a trifluoroacetate salt suitable for biochemical studies.
  • Molecular weight 572.53 g·mol⁻¹; chemical formula C28H27F3N4O6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.