Learn More
Medchemexpress LLC HY-12684 10mg , CH-223191 CAS:301326-22-7 Purity:>98%
SDP
Catalog No. 502260655
Encompass_Preferred
HY-12684 10mg CH-223191 CAS: 301326-22-7 CH-223191 is a potent and specific antagonist of aryl hydrocarbon receptor (AhR).CH-223191 inhibits TCDD-mediated nuclear translocation and DNA binding of AhR, and inhibits TCDD-induced luciferase activity with an IC50 of 0.03 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0655 Supplier Medchemexpress LLC Supplier No. HY1268410MG
HY-12684 10mg CH-223191 CAS: 301326-22-7 CH-223191 is a potent and specific antagonist of aryl hydrocarbon receptor (AhR).CH-223191 inhibits TCDD-mediated nuclear translocation and DNA binding of AhR, and inhibits TCDD-induced luciferase activity with an IC50 of 0.03 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.