Learn More
Selleck Chemical LLC CH-223191
SDP
Catalog No. 5000002997
Encompass_Preferred
CH-223191 is a potent and specific aryl hydrocarbon receptor (AhR) antagonist with IC50 of 30 nM
Catalog No. 50-000-02997 Supplier Selleck Chemical LLC Supplier No. S7711200MG
CH-223191 is a potent and specific aryl hydrocarbon receptor (AhR) antagonist with IC50 of 30 nM