Learn More
TARGETMOL CHEMICALS INC CH-223191 25MG
SDP
Catalog No. 502580803
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CH-223191 is a potent and specific antagonist for aryl hydrocarbon receptor (AhR). purity: 99%
Catalog No. 50-258-0803 Supplier TARGETMOL CHEMICALS INC Supplier No. T244825MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CH-223191 is a potent and specific antagonist for aryl hydrocarbon receptor (AhR). purity: 99%