Learn More
Waters Corp Waters Tubing Kit, MP35N, for ACQUITY UPLC Column Heater-30A
Catalog No. 502039287 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit includes various MP35N tubes for use with the ACQUITY UPLC Column Heater-30A. These tubes serve purposes such as APH for lower port with 30 cm columns, pre-heater to column, column to detector, and an alternate column to detector tube.
  • APH for use in lower port with 30 cm columns.
  • Pre-heater to column tube for APH to 30 cm column with guard, short fitting.
  • Column to detector tube.
  • Alternate column to detector tube.
  • Reusable pre-heater to column tube for APH to 30 cm column.
Catalog No. 50-203-9287 Supplier Waters Corp Supplier No. 205001828
This tubing kit includes various MP35N tubes for use with the ACQUITY UPLC Column Heater-30A. These tubes serve purposes such as APH for lower port with 30 cm columns, pre-heater to column, column to detector, and an alternate column to detector tube.
  • APH for use in lower port with 30 cm columns.
  • Pre-heater to column tube for APH to 30 cm column with guard, short fitting.
  • Column to detector tube.
  • Alternate column to detector tube.
  • Reusable pre-heater to column tube for APH to 30 cm column.