Learn More
Bioworld CH ACTIVATED SEPAROPOREAG
Supplier:  Bioworld 201810851
Encompass
CH ACTIVATED SEPAROPORE (AGAROSE) 4B: Activated CH Separopore 4B is a pre-activated medieum for covalent immobilization of proteins and other ligands containing primary amino groups. Technical Specifications: Group to be coupled: -NH2 ...
Catalog No. 50-197-889