Learn More
Immunology Consultants FITC Conjugated Chicken anti-DYKDDDDK (FLAG) Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants CFLG45F
Encompass_Preferred
Chickens were immunized with DYKDDDDK peptide conjugated with KLH. Eggs were collected and the Ig fraction prepared. Antibodies were immunoaffinity purified using the peptide immobilized on a solid phase matrix then conjugated with fluorescein isothiocyanate (FITC). Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. FITC/Protein Ratio = 3.0 –6.0. This antibody will react with DYKDDDDK as determined by ELISA and IEP techniques. It is suitable for blotting, ELISA and IF applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2072