Learn More
Enzo Life Sciences Chaetocin (200µg). CAS: 28097-03-2
SDP
Supplier:  Enzo Life Sciences BMLGR3490200
Encompass_Preferred
Chaetocin, an antitumor antibiotic, is a thiodioxopiperazine natural product produced by Chaetomium species. Specific inhibitor of the lysine-specific methyltransferase SU (VAR)3-9 both in vitro and in vivo. It displays potent antimyeloma activity in IL-6-dependent myeloma cell lines. Its antimyeloma activity appears to be due to induction of oxidative stress and consequent apoptosis. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml), 100% ethanol (25mg/ml), dimethyl formamide or methanol. Poorly soluble in water. Long Term Storage: .
Catalog No. 50-201-1002