Learn More
Enzo Life Sciences Chaetomellic acid A (5mg). CAS: 148796-51-4
SDP
Supplier:  Enzo Life Sciences BMLG2290005
Encompass_Preferred
Potent inhibitor of farnesyltransferase in isolated enzyme assays (IC50=55nM) but is inactive in whole cells. Inhibition of farnesyltransferase is selective over geranylgeranyltransferase I and II (IC50=92µM and 34µM respectively). Purity: ≥98% (TLC). Solubility: Soluble in warm water (2mg/ml, slightly cloudy). Long Term Storage: -20°C.
Catalog No. 50-201-0861