Learn More
Aviva Systems Biology Corp CH25H Antibody - C-terminal region (ARP65536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CH25H. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human CH25H. Alternate names: C25HApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-514