Learn More
Medchemexpress LLC CH2COOH-PEG3-CH2COOH | 32775-08-9 | 99.1% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1226965G
Encompass_Preferred
CH2COOH-PEG3-CH2COOH is a PEG-based linker bearing two terminal carboxylic acid groups used as a building block in the chemical synthesis of Proteolysis Targeting Chimeras (PROTACs) and other conjugates for research.
  • Provides two terminal carboxylic acids for covalent or amide coupling
  • Peg3 spacer enhances solubility and flexibility in conjugates
  • High listed purity suitable for research use
  • Molecular weight 266.25; chemical formula C10H18O8
  • Available in small-scale packages for synthetic work
Catalog No. 50-002-30154