Learn More
Medchemexpress LLC CH3OCO-D-CHA-Gly-Arg-pNA acetate | 80895-10-9 | C27H42N8O9 | 100 MG
SDP
Catalog No. 5000376332
Encompass_Preferred
CH3OCO-D-CHA-Gly-Arg-pNA acetate is a chromogenic substrate for factor Xa. Intended for research use only, this product has a purity of 99.24% and appears as a white to off-white solid.
  • Chromogenic substrate for factor Xa
  • For research use only
  • 99.24% purity
  • White to off-white solid appearance
Catalog No. 50-003-76332 Supplier Medchemexpress LLC Supplier No. HY153829100MG
CH3OCO-D-CHA-Gly-Arg-pNA acetate is a chromogenic substrate for factor Xa. Intended for research use only, this product has a purity of 99.24% and appears as a white to off-white solid.
  • Chromogenic substrate for factor Xa
  • For research use only
  • 99.24% purity
  • White to off-white solid appearance