Learn More
Medchemexpress LLC HY-15466 5mg Medchemexpress, CH5132799 CAS:1007207-67-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY154665MG
Encompass_Preferred
Medchemexpress, HY-15466 5mg CH5132799 CAS:1007207-67-1 CH5132799 is a selective class I PI3K inhibitor. CH5132799 inhibits class I PI3Ks, particularly PI3Kα, with an IC50 of 14 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0634