Learn More
Apexbio Technology LLC CH5138303 959763-06-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063886
Encompass_Preferred
CH5138303 (CAS 959763-06-5) is a potent inhibitor targeting Heat Shock Protein 90 (HSP90) exhibiting strong binding affinity with a Kd of 0 48 nM HSP90 a molecular chaperone regulates protein folding stability and degradation and maintains oncogenic proteins crucial to tumor cell proliferation CH5138303 effectively reduces protein levels and phosphorylation of HSP90 client proteins including Raf1 AKT EGFR and HER2 in NCI-N87 cancer cells In animal studies using human gastric cancer NCI-N87 xenografts oral administration of CH5138303 demonstrates significant antitumor activity achieving a tumor growth inhibition (TGI) rate of 136% at an ED of 3 9 mg/kg
Catalog No. 50-000-63886 Supplier Apexbio Technology LLC Supplier No. B45715.1
 Restricted Item
Add to Cart
Add to Cart
CH5138303 (CAS 959763-06-5) is a potent inhibitor targeting Heat Shock Protein 90 (HSP90) exhibiting strong binding affinity with a Kd of 0 48 nM HSP90 a molecular chaperone regulates protein folding stability and degradation and maintains oncogenic proteins crucial to tumor cell proliferation CH5138303 effectively reduces protein levels and phosphorylation of HSP90 client proteins including Raf1 AKT EGFR and HER2 in NCI-N87 cancer cells In animal studies using human gastric cancer NCI-N87 xenografts oral administration of CH5138303 demonstrates significant antitumor activity achieving a tumor growth inhibition (TGI) rate of 136% at an ED of 3 9 mg/kg