Learn More
Selleck Chemical LLC CH5183284 (Debio-1347) 5mg 1265229-25-1 FF284
SDP
Supplier:  Selleck Chemical LLC S76655MG
Encompass_Preferred
CH5183284 (Debio-1347, FF284) is a selective and orally available FGFR inhibitor with IC50 of 9.3 nM, 7.6 nM, 22 nM, and 290 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1326501