Learn More
Aviva Systems Biology Corp CHAC1 Antibody - C-terminal region (ARP42623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHAC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CHAC1. Alternate names: MGC4504ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53345