Learn More
Aviva Systems Biology Corp CHAD Antibody - middle region (ARP51926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHAD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHAD. Alternate names: SLRR4AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-60646