Learn More
ABclonal Technology CHAD Rabbit pAb
SDP
Catalog No. 5000105612
Encompass_Preferred
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.
Catalog No. 50-001-05612 Supplier ABclonal Technology Supplier No. A14985500UL
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.