Learn More
Medchemexpress LLC HY-N2019 5mg , Chaetocin CAS:28097-03-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20195MG
Encompass_Preferred
HY-N2019 5mg Chaetocin CAS: 28097-03-2 Chaetocin is a specific inhibitor of the histone methyltransferase (HMT) SU(VAR)3-9 with an IC50 of 0.6 μM for SU(VAR)3-9. It also inhibits thioredoxin reductase (TrxR) with an IC50 of 4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0316