Learn More
Abcam Chaetocin, histone lysine methyltransferase inhibitor, 200UG
Catalog No. 502705644
Encompass_Preferred
MW 696.8 Da, Purity >98%. Specific inhibitor of the lysine-specific methyltransferase SU (VAR)3-9 both in vitro and in vivo. It displays potent antimyeloma activity in IL-6-dependent myeloma cell lines. Its antimyeloma activity appears to be due to induction of oxidative stress and consequent apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5644 Supplier Abcam Supplier No. AB144534200UG
Add to Cart
Add to Cart
MW 696.8 Da, Purity >98%. Specific inhibitor of the lysine-specific methyltransferase SU (VAR)3-9 both in vitro and in vivo. It displays potent antimyeloma activity in IL-6-dependent myeloma cell lines. Its antimyeloma activity appears to be due to induction of oxidative stress and consequent apoptosis.
The product is subject to the following: Abcam Restricted Use Statement