Learn More
AdipoGen Chaetoglobosin A
SDP
Catalog No. 502562106
Encompass_Preferred
Chemical. CAS 50335-03-0. Formula C32H36N2O5. MW 528.7. Isolated from Chaetomium sp. Antibiotic. Cytochalasan analog with anti-fungal activity. Phytotoxic, nematicidal and anti-bacterial activity. Exhibits cytotoxic effects against human cancer cell lines. Induces apoptosis in lymphocytic leukemia cells by targeting the cytoskeleton. Enhances fibrinolytic activity of bovine aortic endothelial cells. Mycotoxin.
Catalog No. 50-256-2106 Supplier AdipoGen Supplier No. BVT0092M005
Chemical. CAS 50335-03-0. Formula C32H36N2O5. MW 528.7. Isolated from Chaetomium sp. Antibiotic. Cytochalasan analog with anti-fungal activity. Phytotoxic, nematicidal and anti-bacterial activity. Exhibits cytotoxic effects against human cancer cell lines. Induces apoptosis in lymphocytic leukemia cells by targeting the cytoskeleton. Enhances fibrinolytic activity of bovine aortic endothelial cells. Mycotoxin.