Learn More
AdipoGen Chaetoglobosin C
SDP
Catalog No. 502561901
Encompass_Preferred
Chemical. CAS 50645-76-6. Formula C32H36N2O5. MW 528.7. Isolated from Chaetomium sp. Cytochalasan mycotoxin, derivative of chaetoglobosin A Prod. No. BVT-0092 http//www.adipogen.com/bvt-0092/chaetoglobosin-a.html . Phytotoxic and antifungal compound. Moderate cytotoxic compound.
Catalog No. 50-256-1901 Supplier AdipoGen Supplier No. BVT0460M001
Chemical. CAS 50645-76-6. Formula C32H36N2O5. MW 528.7. Isolated from Chaetomium sp. Cytochalasan mycotoxin, derivative of chaetoglobosin A Prod. No. BVT-0092 http//www.adipogen.com/bvt-0092/chaetoglobosin-a.html . Phytotoxic and antifungal compound. Moderate cytotoxic compound.