Learn More
AdipoGen Chaetoglobosin F
SDP
Catalog No. 502561900
Encompass_Preferred
Chemical. CAS 55945-75-0. Formula C32H38N2O5. MW 530.7. Isolated from Chaetomium sp. Cytochalasan mycotoxin, derivative of chaetoglobosin A Prod. No. BVT-0092 http//www.adipogen.com/bvt-0092/chaetoglobosin-a.html . Phytotoxic. Cytotoxic against HCT116 human colon cancer cells. Antibacterial Ps. aeruginosa. Anti-inflammatory and immunomodulatory compound.
Catalog No. 50-256-1900 Supplier AdipoGen Supplier No. BVT0461M001
Chemical. CAS 55945-75-0. Formula C32H38N2O5. MW 530.7. Isolated from Chaetomium sp. Cytochalasan mycotoxin, derivative of chaetoglobosin A Prod. No. BVT-0092 http//www.adipogen.com/bvt-0092/chaetoglobosin-a.html . Phytotoxic. Cytotoxic against HCT116 human colon cancer cells. Antibacterial Ps. aeruginosa. Anti-inflammatory and immunomodulatory compound.