Learn More
AdipoGen Chaetoviridin A
SDP
Catalog No. 502561938
Encompass_Preferred
Chemical. CAS 128252-98-2. Formula C23H25ClO6. MW 432.9. Isolated from Chaetomium sp. Azaphilone antibiotic. Antifungal compound phytopathogenic fungi. Cholesteryl ester transfer protein CETP inhibitor. Improves blood lipid levels. Monoamine oxidase inhibitor.
Catalog No. 50-256-1938 Supplier AdipoGen Supplier No. BVT0419M005
Chemical. CAS 128252-98-2. Formula C23H25ClO6. MW 432.9. Isolated from Chaetomium sp. Azaphilone antibiotic. Antifungal compound phytopathogenic fungi. Cholesteryl ester transfer protein CETP inhibitor. Improves blood lipid levels. Monoamine oxidase inhibitor.