Learn More
Aviva Systems Biology Corp CHAF1B Antibody - C-terminal region (ARP45827_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45827_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CHAF1B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CHAF1B. Alternate names: CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59148