Learn More
Amsbio LLC ChagoK1FFPE Cell Pellet Slide, 5 slides
Catalog No. NC3779426
Encompass
Size: 5 slides. Species: Human. Cell Line: ChagoK1. Tissue Type: Lung. Disease: Cancer. Format: FFPE. Storage: 2-8oC
Catalog No. NC3779426 Supplier Amsbio LLC Supplier No. 302044205PK
Size: 5 slides. Species: Human. Cell Line: ChagoK1. Tissue Type: Lung. Disease: Cancer. Format: FFPE. Storage: 2-8oC