Learn More
Cole-Parmer Cole-Parmer Chain Clamp, Large, 11" (280 mm) Dia, Nickel-Plated Zinc
Catalog No. NC1612084 Shop All Cole-Parmer Products
Encompass_Preferred
Chain clamps have a quick slip-on-chain connection and a large easy-to-turn adjustment knob. Holds circular and irregularly shaped apparatus firmly. Clamps include integral connector and slip-on vinyl sleeve. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
Catalog No. NC1612084 Supplier Cole-Parmer Supplier No. 0804734
Chain clamps have a quick slip-on-chain connection and a large easy-to-turn adjustment knob. Holds circular and irregularly shaped apparatus firmly. Clamps include integral connector and slip-on vinyl sleeve. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.