Learn More
MSC Value Collection Offset Link, Carbon Steel, ANSI 35, Single Strand, 10/Pk.
Catalog No. 5000514625
Encompass
This offset link is designed for use with single strand roller chains, providing a reliable connection for mechanical drive and conveyor systems. Constructed from durable carbon steel, this component ensures strength and longevity in industrial applications.
  • Compatible with ANSI number 35 roller chains
  • Manufactured from robust carbon steel for high performance
  • Supplied as a 10 pack for convenient maintenance and repair
  • Engineered for secure integration into power transmission assemblies
Catalog No. 50-005-14625 Supplier MSC Supplier No. 53601936
This offset link is designed for use with single strand roller chains, providing a reliable connection for mechanical drive and conveyor systems. Constructed from durable carbon steel, this component ensures strength and longevity in industrial applications.
  • Compatible with ANSI number 35 roller chains
  • Manufactured from robust carbon steel for high performance
  • Supplied as a 10 pack for convenient maintenance and repair
  • Engineered for secure integration into power transmission assemblies