Learn More
Bevco Precision Manufacturing Co Everlast Tall Height ESD/ISO 4 Cleanroom Black Polyurethane Chair, Articulating Seat & Back Tilt; Chrome Tubular Steel Base w/Welded Footring
Catalog No. NC2022969 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
BEVCO's Everlast ESD/ISO 4 Cleanroom chairs are designed for outstanding comfort and long-lasting use in highly technical environments. The black ESD polyurethane seat and back are static dissipative and are impervious to water, oil, and most chemicals, puncture resistant and easy to clean. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. The waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 24" - 29". This chair comes with a sturdy 22" diameter chrome tubular steel base with brass drag chain and welded footring. Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.
Catalog No. NC2022969 Supplier Bevco Precision Manufacturing Co Supplier No. 7610E1
BEVCO's Everlast ESD/ISO 4 Cleanroom chairs are designed for outstanding comfort and long-lasting use in highly technical environments. The black ESD polyurethane seat and back are static dissipative and are impervious to water, oil, and most chemicals, puncture resistant and easy to clean. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. The waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 24" - 29". This chair comes with a sturdy 22" diameter chrome tubular steel base with brass drag chain and welded footring. Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.