Learn More
Eurofins Discoverx ChaK1, active, 10 µg. (14-961)
Supplier:  Eurofins Discoverx 14961
Encompass
N-terminal 6xHis-tagged recombinant, human ChaK1 amino acids 1180-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9490