Learn More
Sino Biological Recombinant Human TRPM7 Protein (GST Tag), Active
Catalog No. 5000083945
Encompass_Preferred
Recombinant human CHAK1 (TRPM7) (1158-1865) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-000-83945 Supplier Sino Biological Supplier No. T4011G50
Recombinant human CHAK1 (TRPM7) (1158-1865) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.