Learn More
Medchemexpress LLC Chalcogen-containing ionizable cationic lipidoid | 2227214-78-8 | 98.1% | 750.15 g/mol | C44H83N3O6 | 5 MG
SDP
Catalog No. 5000284924
Encompass_Preferred
93-O17O is a chalcogen-containing ionizable cationic lipidoid (C44H83N3O6, MW 750.15 g/mol) used to formulate lipid nanoparticles (LNPs) for nucleic acid delivery research. Supplied as a research-grade, high-purity lipid for formulation, transfection, and preclinical studies. Handle and store according to safety guidance.
  • Used for LNP formulation and mRNA/nucleic acid delivery studies.
  • Chalcogen-containing ionizable cationic structure to facilitate endosomal release.
  • High purity suitable for formulation work (≈98.1%).
  • Available in small-quantity packages for research use.
  • Storage: pure material stable at -20°C; in solvent recommended -80°C for extended storage.
Catalog No. 50-002-84924 Supplier Medchemexpress LLC Supplier No. HYW5981785MG
93-O17O is a chalcogen-containing ionizable cationic lipidoid (C44H83N3O6, MW 750.15 g/mol) used to formulate lipid nanoparticles (LNPs) for nucleic acid delivery research. Supplied as a research-grade, high-purity lipid for formulation, transfection, and preclinical studies. Handle and store according to safety guidance.
  • Used for LNP formulation and mRNA/nucleic acid delivery studies.
  • Chalcogen-containing ionizable cationic structure to facilitate endosomal release.
  • High purity suitable for formulation work (≈98.1%).
  • Available in small-quantity packages for research use.
  • Storage: pure material stable at -20°C; in solvent recommended -80°C for extended storage.