Learn More
AdipoGen Chalcomycin
SDP
Supplier:  AdipoGen BVT0263M001
Encompass_Preferred
Chemical. CAS 20283-48-1. Formula C35H56O14. MW 700.8. Isolated from Streptomyces bikiniensis. 16-membered macrolide antibiotic. Broad spectrum antimicrobial. Antibacterial and antifungal. Protein biosynthesis inhibitor.
Catalog No. 50-256-2026