Learn More
Selleck Chemical LLC Chalcone
SDP
Catalog No. 5000003915
Encompass_Preferred
Chalcone (benzylideneacetophenone phenyl styryl ketone benzalacetophenone -phenylacrylophenone) is an aromatic ketone that forms the central core for a variety of important biological compounds which are known collectively as chalcones with antibacterial antifungal antitumor and anti-inflammatory properties
Catalog No. 50-000-03915 Supplier Selleck Chemical LLC Supplier No. S584525MG
Chalcone (benzylideneacetophenone phenyl styryl ketone benzalacetophenone -phenylacrylophenone) is an aromatic ketone that forms the central core for a variety of important biological compounds which are known collectively as chalcones with antibacterial antifungal antitumor and anti-inflammatory properties