Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chalcone 4 hydrate 1mg | 1202866-96-3 | ≈288.73 g/mol (anhydrous basis); varies with hydration (xH2O) | C16H13ClO3·xH2O | 1 MG
SDP

Catalog No. 5000229327
Encompass_Preferred

Chalcone 4 hydrate (CAS 1202866-96-3) is an anti-parasite small-molecule research reagent reported to inhibit the growth of Babesia and Theileria. The compound is supplied as a hydrated solid and is intended for in vitro parasitology and chemical biology studies; handle and store under recommended conditions.

  • Hydrated solid form (xH2O)
  • Purity 98.0%
  • Molecular formula C16H13ClO3·xH2O
  • Typical molecular weight ~288.73 g/mol (anhydrous basis)
  • Offered in low-quantity research sizes (e.g., 1 mg)
  • Store at 4°C; in solution, store -80°C up to 6 months or -20°C up to 1 month under nitrogen

Catalog No. 50-002-29327 Supplier Medchemexpress LLC Supplier No. HY1155501MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chalcone 4 hydrate (CAS 1202866-96-3) is an anti-parasite small-molecule research reagent reported to inhibit the growth of Babesia and Theileria. The compound is supplied as a hydrated solid and is intended for in vitro parasitology and chemical biology studies; handle and store under recommended conditions.

  • Hydrated solid form (xH2O)
  • Purity 98.0%
  • Molecular formula C16H13ClO3·xH2O
  • Typical molecular weight ~288.73 g/mol (anhydrous basis)
  • Offered in low-quantity research sizes (e.g., 1 mg)
  • Store at 4°C; in solution, store -80°C up to 6 months or -20°C up to 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.