Learn More
TARGETMOL CHEMICALS INC CHALCONE 4 HYDRATE 5MG
SDP
Catalog No. 502581664
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Chalcone 4 hydrate an anti-parasite compound inhibits the growth of Theileria and Babesia. purity: 98%
Catalog No. 50-258-1664 Supplier TARGETMOL CHEMICALS INC Supplier No. T107855MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Chalcone 4 hydrate an anti-parasite compound inhibits the growth of Theileria and Babesia. purity: 98%