Learn More
Medchemexpress LLC Chalcone | 94-41-7 | 100.0% | 50 MG
SDP
Catalog No. 5000232737
Encompass_Preferred
Chalcone is the parent α,β-unsaturated ketone scaffold used widely as a small-molecule research chemical and synthetic building block. Provided as a high-purity solid, it is used in medicinal chemistry, biochemical assays, and the synthesis of chalcone derivatives. Typical identifiers: CAS 94-41-7; formula C15H12O; molecular weight 208.25 g/mol.
  • High purity suitable for research use.
  • Useful building block for synthesis of bioactive derivatives.
  • Compatible with DMSO; available as solution for screening.
  • Supplied with certificate of analysis and safety data for quality and safety.
  • Available in small-scale quantities for discovery research.
Catalog No. 50-002-32737 Supplier Medchemexpress LLC Supplier No. HY12105450MG
Chalcone is the parent α,β-unsaturated ketone scaffold used widely as a small-molecule research chemical and synthetic building block. Provided as a high-purity solid, it is used in medicinal chemistry, biochemical assays, and the synthesis of chalcone derivatives. Typical identifiers: CAS 94-41-7; formula C15H12O; molecular weight 208.25 g/mol.
  • High purity suitable for research use.
  • Useful building block for synthesis of bioactive derivatives.
  • Compatible with DMSO; available as solution for screening.
  • Supplied with certificate of analysis and safety data for quality and safety.
  • Available in small-scale quantities for discovery research.