Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chalcone dibromide | 611-91-6 | MFCD00017861 | 95.0% | 368.06 g·mol⁻1 | C15H12Br2O | 1 ML
SDP

Catalog No. 5000299634
Encompass_Preferred

Chalcone dibromide (CAS 611-91-6) is a dibrominated chalcone derivative supplied as a ready-to-use 10 mM solution in DMSO for research and synthetic applications. It is used as a synthon in the synthesis of bioactive heterocycles and has reported antioxidant and anticancer-related activity; product documentation includes datasheet, COA, and SDS.

  • Supplied as a 10 mM solution in DMSO (1 mL).
  • Molecular formula C15H12Br2O and molecular weight 368.06 g·mol⁻1.
  • Purity 95.0%.
  • Useful synthon for synthesis of pyrazolines, hydroxy pyrazolines, and isoxazoles.
  • Available as a solid (100 mg) and larger sizes by quote.
  • Documentation includes datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-99634 Supplier Medchemexpress LLC Supplier No. HYW02398310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chalcone dibromide (CAS 611-91-6) is a dibrominated chalcone derivative supplied as a ready-to-use 10 mM solution in DMSO for research and synthetic applications. It is used as a synthon in the synthesis of bioactive heterocycles and has reported antioxidant and anticancer-related activity; product documentation includes datasheet, COA, and SDS.

  • Supplied as a 10 mM solution in DMSO (1 mL).
  • Molecular formula C15H12Br2O and molecular weight 368.06 g·mol⁻1.
  • Purity 95.0%.
  • Useful synthon for synthesis of pyrazolines, hydroxy pyrazolines, and isoxazoles.
  • Available as a solid (100 mg) and larger sizes by quote.
  • Documentation includes datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.