Learn More
Medchemexpress LLC Chalcone dibromide | 611-91-6 | 95.0% | 368.06 g/mol | C15H12Br2O | 100 MG
SDP
Catalog No. 5000300454
Encompass_Preferred
Chalcone dibromide is a dibrominated chalcone derivative supplied as a white to off-white solid for research use. It serves as a versatile synthetic building block and bioactive compound in medicinal chemistry and organic synthesis, commonly used to prepare heterocycles and to evaluate biological activity.
  • Dibrominated chalcone useful as a synthetic intermediate.
  • Solid, white to off-white appearance for easy handling.
  • Reported purity of 95.0% for research applications.
  • Available in multiple pack formats including 100 mg and DMSO solutions.
  • Structural identifiers provided (SMILES) to support characterization.
Catalog No. 50-003-00454 Supplier Medchemexpress LLC Supplier No. HYW023983100MG
Chalcone dibromide is a dibrominated chalcone derivative supplied as a white to off-white solid for research use. It serves as a versatile synthetic building block and bioactive compound in medicinal chemistry and organic synthesis, commonly used to prepare heterocycles and to evaluate biological activity.
  • Dibrominated chalcone useful as a synthetic intermediate.
  • Solid, white to off-white appearance for easy handling.
  • Reported purity of 95.0% for research applications.
  • Available in multiple pack formats including 100 mg and DMSO solutions.
  • Structural identifiers provided (SMILES) to support characterization.