Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chalepensin | 13164-03-9 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN83821MG

Encompass_Preferred

Chalepensin is a furanocoumarin compound that functions as a competitive inhibitor of CYP2A6. It also demonstrates inhibitory activity against various other human cytochrome P450 enzymes, including CYP1A1, CYP1A2, CYP2A13, CYP2C9, CYP2D6, CYP2E1, and CYP3A4.

  • Furanocoumarin compound
  • Competitive CYP2A6 inhibitor
  • Inhibits human CYP1A1, CYP1A2, CYP2A13, CYP2C9, CYP2D6, CYP2E1, and CYP3A4
  • Molecular weight: 254.28
  • Formula: C16H14O3
  • Appearance: Solid, white to off-white

Catalog No. 50-004-17454


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.