Learn More
Medchemexpress LLC Chalepensin | 13164-03-9 | 5 MG

Supplier: Medchemexpress LLC HYN83825MG
Chalepensin is a furanocoumarin that acts as a competitive CYP2A6 inhibitor. It also demonstrates inhibitory effects on human CYP1A1, CYP1A2, CYP2A13, CYP2C9, CYP2D6, CYP2E1, and CYP3A4 to different extents. This product is intended for research use only and is not for sale to patients.
- Competitive CYP2A6 inhibitor
- Inhibits human CYP1A1, CYP1A2, CYP2A13, CYP2C9, CYP2D6, CYP2E1, and CYP3A4
- For research use only
- Purity: 98.98%
- Molecular weight: 254.28
- Chemical formula: C16H14O3
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.