Learn More
Caron Products And Services Environmental Chamber, 2-Door, 50 cu. ft., Temp & RH
Catalog No. NC2110203
Encompass_Preferred
50 ft3 (1,416 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
This item is not returnable. View return policy
Catalog No. NC2110203 Supplier Caron Products And Services Supplier No. 7000501
Add to Cart
Add to Cart
Request a Quote
50 ft3 (1,416 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.