Learn More
Med Vet International AM Induction Chamber - Large (20L), J0265M, Each
Catalog No. 5000495830
Encompass_Preferred
Large chamber for cats, small dogs, and rabbits. ? Sealed w/gasket for faster inductions ? Less waste anesthetic gas leakage ? Secure and robust one-piece molded base ? Simple anesthetic connection ? Easy to clean ? No joints ? Can use for euthanasia chamber ? Fresh gas inlet is metal barbed fitting with 5? of smooth vinyl tubing w/proximal 15mm ET connector ? Proximal end has 15mm o.d. connector ? Gas outlet 22mm tubing Dimensions: 9? (23cm)W x 17? (42cm)L x 8? (20cm)H
Catalog No. 50-004-95830 Supplier Med Vet International Supplier No. JOR265M
Large chamber for cats, small dogs, and rabbits. ? Sealed w/gasket for faster inductions ? Less waste anesthetic gas leakage ? Secure and robust one-piece molded base ? Simple anesthetic connection ? Easy to clean ? No joints ? Can use for euthanasia chamber ? Fresh gas inlet is metal barbed fitting with 5? of smooth vinyl tubing w/proximal 15mm ET connector ? Proximal end has 15mm o.d. connector ? Gas outlet 22mm tubing Dimensions: 9? (23cm)W x 17? (42cm)L x 8? (20cm)H