Learn More
Med Vet International AM Induction Chamber - Medium (6.0L), J0265N, Each
Catalog No. 5000495881
Encompass_Preferred
Middle size for rodents, birds, and pocket pets. ? Sealed round corner and side clip ? Aestheticize several patients at once ? Cap unscrews to remove patient ? Simple anesthetic connection ? Easy to clean ? No joints ? Can use for euthanasia chamber ? Fresh gas inlet is metal barbed fitting with 5? of smooth vinyl tubing w/proximal 15mm ET connector ? Proximal end has 15mm o.d. connector ? Gas outlet 22mm tubing Dimensions: 7? W x 10? L x 5? H
Catalog No. 50-004-95881 Supplier Med Vet International Supplier No. JOR265N
Middle size for rodents, birds, and pocket pets. ? Sealed round corner and side clip ? Aestheticize several patients at once ? Cap unscrews to remove patient ? Simple anesthetic connection ? Easy to clean ? No joints ? Can use for euthanasia chamber ? Fresh gas inlet is metal barbed fitting with 5? of smooth vinyl tubing w/proximal 15mm ET connector ? Proximal end has 15mm o.d. connector ? Gas outlet 22mm tubing Dimensions: 7? W x 10? L x 5? H